Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Factor X (FITC), Polyclonal, Invitrogen™

Manufacturer:  Thermo Scientific PA174114

Catalog No. PIPA174114

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Host Species: Sheep
Isotype: IgG
Formulation: 100µg of Affinity chromatography purified antibody in PBS buffer, with BSA 1mg/mL; pH 7.4.
Immunogen: Factor Xa (active site blocked) from human factor X.

PA1-74114 detects Factor X from human samples. PA1-74114 has been successfully used in ELISA applications. The PA1-74114 immunogen is Factor Xa (active site blocked) from human factor X.

Specifications

Specifications

Factor X (FITC)
Purified
Factor Xa (active site blocked) from human factor X.
100μg
Primary
FITC
Sheep
IgG
Store at 4°C
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit