Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129169

Catalog No. PA129169

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-29171 detects FRA2 from human samples. PA1-29171 has been successfully used in Western blot applications. The PA1-29171 immunogen is: Synthetic peptide corresponding to the C-terminus of human Fra2 protein.

FAS-associated death domain containing protein (FADD) or MORT1 is a cytoplasmic protein which interacts with FAS to initiate apoptosis.
Specifications

Specifications

FADD
Polyclonal
Unconjugated
FADD
Liquid
FADD
Synthetic peptide corresponding to the N-terminus of human FADD protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
Q13158, Q61160
FAS-associated death domain-containing protein, GIG3, MGC8528, MORT1
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
14082, 266610, 8772
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit