Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-FADS1 Clone: 2D9,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ ST1579100UG
Description
Specifically detects FADS1 Clone: 2D9 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting
FADS1 is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs.Specifications
FADS1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin) | |
2D9 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids 1-100 of human FADS1, expressed as a GST fusion protein | |
RUO | |
Recognizes the ∽50-55kDa FADS1 protein in HeLa cells and human skeletal muscle tissue. | |
−20°C | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction