Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAK Mouse anti-Avian, Human, Mouse, Rat, Clone: 2A7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05182
Description
Anti-FAK Antibody, clone 2A7 detects level of FAK and has been published and validated for use in IC and immunoprecipitation.Specifications
FAK | |
Monoclonal | |
Unconjugated | |
NM_005607.3; NM_153831.2 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunocytochemistry, Immunoprecipitation | |
2A7 | |
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in Purified immunoglobulin in 10mM PBS, pH 7.4 containing no preservatives. | |
PTK2, pp125FAK, FAK, FADK, FAK1 | |
Mixture of affinity-purified tyrosine phosphoproteins from chick embryo fibroblasts expressing p125FAK. | |
200 μg | |
Avian, Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only