anti-FALDH, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 311700020.1MG

 View more versions of this product

Catalog No. 31-170-00201

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Disclaimers:

For Research Use Only


    Specifications

    Format Affinity Purified
    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Gene Accession No. P51648
    Applications ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blotting
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Gene ID (Entrez) 224
    Test Specificity This product is specific for Human FALDH.
    Antigen FALDH
    Host Species Rabbit
    Immunogen This antibody is specific for the N Terminus Region of the target protein.
    Quantity 0.1mg
    Regulatory Status RUO
    Conjugate Unlabeled
    Cross Reactivity Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit