Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FAM126A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132914

Catalog No. NBP132914


Add to cart

Description

Description

FAM126A Polyclonal antibody specifically detects FAM126A in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

FAM126A
1.0mg/mL
Western Blot 1:1000-1:10000
Q9BYI3
Down-regulated by CTNNB1 protein A, DRCTNNB1Aa, family with sequence similarity 126, member A, HLD5, hyccin, Protein FAM126A
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Rat(90%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
FAM126A
Recombinant protein encompassing a sequence within the center region of human FAM126A. The exact sequence is proprietary.
57.6 kDa
100 ul
Signal Transduction
Primary
84668
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only