Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM151A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525076

Catalog No. PIPA525076

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

FAM151A
Polyclonal
Unconjugated
FAM151A
Liquid
FAM151A
KLH conjugated synthetic peptide between 547-576 amino acids from the C-terminal region of human FAM151A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8WW52, Q8QZW3
C1orf179, hyporthetical protein MGC27169, protein FAM151A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230579, 338094
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit