Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM155A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523649

Catalog No. PIPA523649

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

FAM155A
Polyclonal
PBS with 0.09% sodium azide
B1AL88
transmembrane protein FAM155A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
728215
Flow Cytometry, Western Blot
Unconjugated
FAM155A
Liquid
FAM155A
KLH conjugated synthetic peptide between 45-72 amino acids from the N-terminal region of human FAM155A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit