Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FAM177A1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178754

Catalog No. NBP178754


Add to cart

Description

Description

FAM177A1 Polyclonal antibody specifically detects FAM177A1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

FAM177A1
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg lysate
NP_001072987.1
C14orf24, chromosome 14 open reading frame 24, DKFZp686J1254, family with sequence similarity 177, member A1, FLJ38854, hypothetical protein LOC283635
Rabbit
IgG
100 ul
Protein Phosphatase
Primary
283635
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
FAM177A1
The immunogen for this product maps to a region between residue 1 and 50 of human Family with Sequence Similarity 177, Member A1 using the numbering given in entry NP_001072987.1 (GeneID 283635).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only