Learn More
anti-FAM18B, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131988
Catalog No. NBP131988
Add to cart
Description
Description
FAM18B Polyclonal antibody specifically detects FAM18B in Human samples. It is validated for Western Blot.
Specifications
Specifications
FAM18B
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
TVP23B
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human FAM18B. The exact sequence is proprietary.
24 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CGI-148, FAM18B, family with sequence similarity 18, member B, family with sequence similarity 18, member B1, FLJ46240, hypothetical protein LOC51030, NPD008, YDR084C
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51030
Human