Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM26E Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525712

Catalog No. PIPA525712


Add to cart

Specifications

Specifications

FAM26E
Polyclonal
Unconjugated
CALHM5
Liquid
CALHM5, Fam26e
KLH conjugated synthetic peptide between 214-243 amino acids from the C-terminal region of human FAM26E
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8N5C1, Q8R100
C6orf188, dJ493F7.3, protein FAM26E
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
103511, 254228
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit