Learn More
FAM59A Rabbit anti-Equine, Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523255
Catalog No. PIPA523255
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
FAM59A is a recently identified intracellular protein belonging to the FAM59 family. It contains one SAM (sterile alpha motif) domain. Alternative splicing result shows that it has three isoforms. The function of FAM59A is yet unknown.  It is widely expressed in many tissues. 
Specifications
Specifications
FAM59A
Polyclonal
Unconjugated
GAREM1
Liquid
GAREM1, LOC100066443
A portion of amino acids 1-50 of human FAM59A was used as the immunogen for the antibody.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Equine, Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9H706
C18orf11, GAREM, Gm944, Grb2-associated and regulator of Erk/MAPK, protein FAM59A
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
100066443, 64762