Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM78B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524185

Catalog No. PIPA524185

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The exact function of FAM78B remains unknown.
Specifications

Specifications

FAM78B
Polyclonal
PBS with 0.09% sodium azide
Q5VT40, Q8BQN5
protein FAM78B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
149297, 226610
Western Blot
Unconjugated
FAM78B
Liquid
FAM78B
KLH conjugated synthetic peptide between 77-103 amino acids from the central region of human FAM78B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit