Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM92A1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525095
Description
This antibody is predicted to react with bovine based on sequence homology.
The function of this protein has not been specifically defined.Specifications
FAM92A1 | |
Polyclonal | |
FAM92A | |
protein FAM92A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
137392 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
A1XBS5 | |
FAM92A | |
KLH conjugated synthetic peptide between 113-141 amino acids from the central region of human FAM92A1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction