Learn More
FANCA Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522986
Catalog No. PIPA522986
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
FANCA (Fanconi anemia group A protein) is a DNA repair protein. It may function as a cell cycle checkpoint or in postreplication repair. FANCA may be important for the maintece of chromosome stability and reparation of interstrand DNA cross linkages. Defects in this protein result in a disease state, Fanconi anemia, an autosomal recessive disorder. Fanconi anemia results in many congenital malformations and predisposition to maligt formations.
Specifications
Specifications
FANCA
Polyclonal
whole serum with no preservative
O15360
FA, FA-H, FA1, FAA, FACA, FAH, FANCH, Fanconi anemia group A protein, Fanconi anemia, complementation group H, Fanconi anemia, type 1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
FANCA
Liquid
FANCA
Two synthetic peptides made to a region of human FANCA (within amino acids 930-1100 and 1390-1455)
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2175