Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fanconi anemia, complementation group C, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human FANCC protein.

Manufacturer:  Abnova Corporation H00002176B01

Catalog No. 89-005-344


Add to cart

Description

Description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group C. [provided by RefSeq

Sequence: MAQDSVDLSCDYQFWMQKLSVWDQASTLETQQDTCLHVAQFQEFLRKMYEALKEMDSNTVIERFPTIGQLLAKACWNPFILAYDESQKILIWCLCCLINKEPQNSGQSKLNSWIQGVLSHILSALRFDKEVALFTQGLGYAPIDYYPGLLKNMVLSLASELRENHLNGFNTQRRMAPERVASLSRVCVPLITLTDVDPLVEALLICHGREPQEILQPEFFEAVNEAILLKKISLPMSAVVCLWLRHLPSLEKAMLHLFEKLISSERNCLRRIECFIKDSSLPQAACHPAIFRVVDEMFRCALLETDGALEIIATIQVFTQCFVEALEKASKQLRFALKTYFPYTSPSLAMVLLQDPQDIPRGHWLQTLKHISELLREAVEDQTHGSCGGPFESWFLFIHFGGWAEMVAEQLLMSAAEPPTALLWLLAFYYGPRDGRQQRAQTMVQVKAVLGHLLAMSRSSSLSAQDLQTVAGQGTDTDLRAPAQQLIRHLLLNFLLWAPGGHTIAWDVITLMAHTAEITHEIIGFLDQTLYRWNRLGIESPRSEKLARELLKELRTQV
Specifications

Specifications

Fanconi anemia, complementation group C
Unconjugated
No additive
NM_000136.2
FA3|FAC|FACC|FLJ14675
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2176
Western Blot
Mouse polyclonal antibody raised against a full-length human FANCC protein.
FANCC
Serum
FANCC
FANCC (NP_000127.2, 1 a.a. ~ 558 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit