Learn More
FANCC Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522987
Catalog No. PIPA522987
Add to cart
Description
Description
Fanconi Anemia Group C Protein is a DNA repair protein. It may function as a cell cycle check point or in post replication repair. FANCC may be important for the maintece of chromosome stability and reparation of interstrand DNA cross linkages. Defects in this protein result in a disease state, Fanconi anemia, an autosomal recessive disorder. Fanconi anemia results in many congenital malformations and predisposition to maligt formations.
Specifications
Specifications
FANCC
Polyclonal
whole serum with no preservative
Q00597
RP11-80I15.2, FA3, FAC, FACC, Fanconi anemia group C protein
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
FANCC
Liquid
FANCC
C-terminal human FANCC fusion protein.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2176