Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522988
Description
FANCC Polyclonal antibody specifically detects FANCC in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
FANCC | |
Polyclonal | |
Unconjugated | |
FANCC | |
FA3; Fac; Facc; FANCC; Fanconi anemia complementation group C; Fanconi anemia group C protein; Fanconi anemia group C protein homolog; Fanconi anemia, complementation group C; FLJ14675; mir-3074-1; Protein FACC; RP11-80I15.2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
Q00597 | |
FANCC | |
N-terminal human FANCC-GST fusion protein. | |
RUO | |
2176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction