Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB1002566
Description
FANCG Polyclonal specifically detects FANCG in Human samples. It is validated for Western Blot, Immunoblotting.Specifications
FANCG | |
Polyclonal | |
Western Blot 1:1000, Immunoblotting reported in scientific literature (PMID 24910428) | |
O15287 | |
FANCG | |
A peptide corresponding to the C-terminus of human FANCG (within amino acids 550-622). [SwissProt# O15287] | |
Unpurified | |
RUO | |
Primary | |
Human. Immunogen sequence has 88% homology to mouse. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoblot | |
Unconjugated | |
Whole antisera with No Preservative | |
DNA repair protein XRCC9, FAG, Fanconi anemia group G protein, Fanconi anemia, complementation group G, Protein FACG, X-ray repair, complementing defective, in Chinese hamster, 9, XRCC9X-ray repair complementing defective repair in Chinese hamster cells 9 | |
Rabbit | |
69 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
2189 | |
Human | |
Antisera |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction