Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRIP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527096

Catalog No. PIPA527096


Add to cart

Description

Description

PA5-27096 targets FANCJ in IF and WB applications and shows reactivity with Human samples. The PA5-27096 immunogen is recombit fragment corresponding to a region within amino acids 52 and 291 of Human FANCJ. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). This gene may be a target of germline cancer-inducing mutations.
Specifications

Specifications

BRIP1
Polyclonal
Unconjugated
BRIP1
Liquid
BRIP1
Recombit fragment corresponding to a region within amino acids 52 and 291 of Human FANCJ
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q9BX63
BACH1, FANCJ, OF, ATP-dependent RNA helicase BRIP1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1/BRCA2-associated helicase 1, Fanconi anemia group J protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
83990
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit