Learn More
FASTK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA521449
Catalog No. PIPA521449
Add to cart
Description
Description
PA5-21449 targets FAST in ICC, IF, IHC (P), and WB applications and shows reactivity with Human, rat and mouse samples. The PA5-21449 immunogen is a recombit fragment corresponding to a region within amino acids 346 and 549 of FAST (Uniprot IDNo.Q14296). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Specifications
FASTK
Polyclonal
Unconjugated
FASTK
Liquid
FASTK
Recombit fragment corresponding to a region within amino acids 346 and 549 of FAST (Uniprot IDNo.Q14296)
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
6.56 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q14296, Q9JIX9
FAST, FAST kinase, fas-activated serine/threonine kinase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10922, 296741, 66587