Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529635

Catalog No. PIPA529635


Add to cart

Description

Description

PA5-29635 targets Fast skeletal myosin light chain 1 in IF and WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-29635 immunogen is recombit protein fragment corresponding to a region within amino acids 1 and 126 of Human Fast skeletal myosin light chain 1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.
Specifications

Specifications

MYL1
Polyclonal
Unconjugated
MYL1
Liquid
MYL1
Recombit protein fragment corresponding to a region within amino acids 1 and 126 of Human Fast skeletal myosin light chain 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
0.59 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P05976, P05977, P02600
MLC1F, MLC3F, A1 catalytic, A2 catalytic, MLC1/MLC3, MLC1F/MLC3F, myosin light chain 1/3, skeletal muscle isoform, myosin light chain A1/A2, myosin light chain alkali 1/2, myosin, light polypeptide 1, alkali; skeletal, fast
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17901, 4632, 56781
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit