Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCS Anti-Fatigue Rubber Mat

Consist of hollow cylinders that offer stable footing. Mat is able to remove electrostatic charges when they are grounded.

Manufacturer:  SCS 9900

Catalog No. 18-688-342


Add to cart

Description

Description

  • Static Control Anti-Fatigue Mats enable workers to stand comfortably for long periods while minimizing the fatigue associated with standing work.
  • Reversible Conductive Rubber Floor Matting
    Provides a flooring surface that does not generate a static charge and will act as part of the path to ground for an operator wearing ESD footwear
  • Rtt: < 1 x 106 ohms per ANSI/ESD 7.1 and ESD TR53
    Meets the required limits of ANSI/ESD S20.20 and is suitable flooring component in personnel grounding Footwear/Flooring System (Ref: STM97.1 and STM97.2). The resistance range of the surface layer remains low enough to act as a path to ground with the normal buildup of dirt and grime on the mat.
  • Low Profile
    Reduces trip hazards and designed for use in high traffic areas and good for use under chairs and carts.
  • Made in the United States of America
    This item is stocked in Sanford, NC
Specifications

Specifications

3 x 5 ft.
Rubber
0.600 in.
3 ft.
0.91 m
5 ft.
Black
1.52 m
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit