Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC27A1 Mouse anti-Human, Clone: 308420, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524022

Catalog No. PIMA524022


Add to cart

Description

Description

This antibody detects human FATP1 in cell transfectants but not non-transfected parent cells in flow cytometry. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

SLC27A1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q6PCB7
SLC27A1, ACSVL5FATP-1, EC 6.2.1, EC 6.2.1.-, FATP1Solute carrier family 27 member 1, FATPlong-chain fatty acid transport protein 1, Fatty acid transport protein 1, FLJ00336, MGC71751, solute carrier family 27 (fatty acid transporter), member 1
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
376497
Flow Cytometry, Immunocytochemistry, Immunofluorescence
308420
Unconjugated
SLC27A1
Lyophilized
SLC27A1
NS0 mouse myeloma cell line transfected with human FATP1 Met1-Leu646
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit