Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FA2H Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112758

Catalog No. 01-674-880


Add to cart

Description

Description

A suggested positive control for this product is pancreas lysate.

2-Hydroxysphingolipids are a subset of sphingolipids containing 2- hydroxy fatty acids. The 2-hydroxylation occurs during de novo ceramide synthesis and is catalyzed by fatty acid 2-hydroxylase (also known as fatty acid alpha-hydroxylase).
Specifications

Specifications

FA2H
Polyclonal
PBS with 0.08% sodium azide
Q7L5A8
FAAH, FAH1, FAXDC1, FLJ25287, SCS7
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79152
Western Blot
Unconjugated
FA2H
Liquid
FA2H
Synthetic peptide corresponding to human fatty acid 2-hydrolase (FA2H).
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit