Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO44 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA14348
Description
PA1-4348 detects FBG3 from human samples. PA1-4348 has been successfully used in Western blot procedures. By Western blot this antibody detects a ∽31 kDa protein representing FBG3. PA1-4348 immunizing peptide corresponds to amino acid residues 103-119 from human FBG3 Protein.
FBXO44 Polyclonal antibody specifically detects FBXO44 in Human samples. It is validated for Western Blot.Specifications
FBXO44 | |
Polyclonal | |
Unconjugated | |
FBXO44 | |
5730411K09; AV001623; DKFZp781J0852; F Box Gama 3; FBG3; F-box gene 3; F-box only protein 44; F-box only protein 6a; F-box protein 44; F-box protein FBX30; F-box/G-domain protein 3; FBX30; Fbx44; FBX6A; FBXO44; FBXO6A; MGC14140; RGD1562463; RP3-330O12.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9H4M3 | |
FBXO44 | |
Synthetic Peptide: E(103) W K V E D L S R D Q R K E F P N(119) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction