Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14350

Catalog No. PA14350


Add to cart

Description

Description

PA1-4350 detects the FBG5 protein in human samples. PA1-4350 has been successfully used in Western blot procedures. The PA1-4350 immunizing peptide corresponds to amino acid residues 163-175 of the human FBG5 protein. There is 76% sequence identity to mouse.

F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
Specifications

Specifications

FBXO27
Polyclonal
Unconjugated
FBXO27
Liquid
FBXO27
Synthetic peptide: K(163) K Q V L D L E E E G L W(175)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8NI29
F Box Gama 5, F-box only protein 27, F-box/G-domain protein 5, FBXO27
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
126433
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit