Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL12 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521621

Catalog No. PIPA521621


Add to cart

Description

Description

PA5-21621 targets FBXL12 in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-21621 immunogen is a synthetic peptide corresponding to a region within amino acids 265 and 326 of FBXL12 (Uniprot IDNo.Q9NXK8). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

FBXL12
Polyclonal
Unconjugated
FBXL12
Liquid
FBXL12
Synthetic peptide corresponding to a region within amino acids 265 and 326 of FBXL12 (Uniprot IDNo.Q9NXK8)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
Q9NXK8, Q9EPX5
Fbl12, F-box protein FBL12, F-box/LRR-repeat protein 12
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
30843, 54850
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit