Learn More
F-box and leucine-rich repeat protein 18, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FBXL18 protein.
Manufacturer: Abnova Corporation H00080028B01
Description
Members of the F-box protein family, such as FBXL18, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MHAVPRGFGKKVRVGVQSCPPLLGPGVPPALLRVLVSAEEPALPGTPRADWVQLLLRHAPQRARHPQLAPTLQPRTECRGLGGGRHRPAGLPAAPDARTAAQRPYGLRAGQYRPAVPAVAVPVAGQPGHDGEGGVHARALRHVEALQAAEGPQAGAALLQRQRPVLPGAEPVPLAAAPVPGLSQRHPPARCRAGLHGSLPAGCHVPPVHRGVPRHLQEPAAVASPQLPGRAARVKRRHLPSAPRGPDRRHPGRPPGAPGSpecifications
F-box and leucine-rich repeat protein 18 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human FBXL18 protein. | |
FBXL18 | |
FLJ10776/FLJ11467/FLJ26934/FLJ38075/FLJ41541/Fbl18 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
No additive | |
BC013435.1 | |
FBXL18 | |
FBXL18 (AAH13435.1, 1 a.a. ∼ 259 a.a) full-length human protein. | |
RUO | |
Yes | |
80028 | |
Serum |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.