Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216445
Description
FBXO16 Polyclonal specifically detects FBXO16 in Mouse samples. It is validated for Western Blot.Specifications
FBXO16 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
F-box only protein 16, F-box protein 16, FBX16MGC125925, MGC125923, MGC125924 | |
Rabbit | |
35 kDa | |
0.1 mL | |
Primary | |
Mouse. Expected to cross react based on sequence identity: Human (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.61mg/mL | |
Western Blot 1:500-1:3000 | |
Q8IX29 | |
FBXO16 | |
Recombinant protein encompassing a sequence within the center region of human FBXO16. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
157574 | |
Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction