Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522811

Catalog No. PIPA522811


Add to cart

Description

Description

Suggested positive control: antigen standard for FBXO27 (transient overexpression lysate).

Members of the F-box protein family, such as FBXO27, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin (CUL1), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications

Specifications

FBXO27
Polyclonal
Unconjugated
FBXO27
Liquid
FBXO27
Synthetic peptide corresponding to residues 163-175 (KKQVLDLEEEGLW) of the human FBG5 protein. 76% identical to mouse.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8NI29, Q6DIA9
FBG5, Fbx27, F-box only protein 27, F-box protein FBG5, F-box/G-domain protein 5
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
126433, 233040, 499114
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit