Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box protein 36 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human FBXO36 protein.

Manufacturer:  Abnova Corporation H00130888B01

Catalog No. 89-008-532


Add to cart

Description

Description

Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MASWLPETLFETVGQGPPPSKDYYQLLVTRSQVIFRWWKISLRSEYRSTKPGEAKETHEDFLENSHLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTNKLQLQRQLRKRKQKYGNLREKQP
Specifications

Specifications

F-box protein 36
Unconjugated
No additive
BC033935.1
FLJ37592|FLJ41090|Fbx36
Mouse
50 uL
Ubiquitin/Proteasome
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human FBXO36 protein.
FBXO36
Serum
FBXO36
FBXO36 (AAH33935.1, 1 a.a. ~ 188 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
130888
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit