Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO39 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524125

Catalog No. PIPA524125


Add to cart

Description

Description

Members of the F-box protein family, such as FBXO39, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications

Specifications

FBXO39
Polyclonal
PBS with 0.09% sodium azide
Q8N4B4, Q5NBU5
Fbx39, F-box only protein 39
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
162517, 628100
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FBXO39
Liquid
FBXO39
KLH conjugated synthetic peptide between 385-414 amino acids from the C-terminal region of human FBXO39
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit