Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522810

Catalog No. PIPA522810


Add to cart

Description

Description

Suggested positive control: antigen standard for FBXO6 (transient overexpression lysate).

The F box, named after cyclin F in which it was originally observed, is an approximately 40-amino acid motif that binds SKP1. F-box proteins are components of modular E3 ubiquitin protein ligases called SCFs (SKP1, cullin, F-box proteins), which function in phosphorylation-dependent ubiquitination.
Specifications

Specifications

FBXO6
Polyclonal
Unconjugated
FBXO6
Liquid
FBXO6
Synthetic peptide corresponding to residues 268-284 (SEAQPGQKHGQEEAAQS) of the human FBG2 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9NRD1
RP3-330O12.3, FBG2, FBS2, FBX6, Fbx6b, F-box only protein 6, F-box protein FBG2, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, F-box/G-domain protein 2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26270
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit