Learn More
FBXW8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524212
Catalog No. PIPA524212
Add to cart
Description
Description
This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains a WD-40 domain, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
Specifications
FBXW8
Polyclonal
Unconjugated
FBXW8
Liquid
FBXW8
KLH conjugated synthetic peptide between 270-299 amino acids from the central region of human FBXW8
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8N3Y1, Q8BIA4
FBW6, FBW8, FBX29, FBXO29, FBXW6, F-box and WD-40 domain protein 8, F-box and WD-40 domain-containing protein 8, F-box only protein 29, F-box/WD repeat-containing protein 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
231672, 26259