Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Fc epsilon RI alpha, Alexa Fluor 488, Clone: 773704, R&D Systems™
Supplier: R&D Systems FAB6678G
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
Fc epsilon RI alpha | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FCER1A | |
Mouse myeloma cell line NS0-derived recombinant human Fc epsilon RI alpha , Val26-Gln205, Accession # P12319 | |
100 Tests | |
Primary | |
Human | |
IgG2b |
Flow Cytometry | |
773704 | |
Flow Cytometry 5 uL/10^6 cells | |
P12319 | |
Mouse | |
Affinity Purified | |
RUO | |
Detects human Fc epsilon RI alpha in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction