Learn More
anti-Fc gamma RIII (CD16), PE, Clone: 245536, R&D Systems™
Manufacturer:  R&D Systems FAB2546P100
 View more versions of this product
Catalog No. FAB2546P100
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
Fc gamma RIII (CD16)
245536
Flow Cytometry 10 uL/10^6 cells
O75015
CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20
Mouse
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Fc gamma RIIA and recombinant mouse Fc gamma RIII is observed.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
FCGR3A
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIA/B (CD16), Thr20-Gln208, Accession # O75015
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2214
Human