Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FceR1 alpha, FITC, clone: CRA2 (AER24), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA15025
Description
Fc epsilonR1 alpha is a subunit of the high affinity receptor of IgE to which it directly binds. Fc epsilonR1 alpha is a tetrameric complex consisting of one alpha, one β and two additional subunits. The latter two are required for signal transduction activity. The Fc epsilonR1 complex plays an important role in triggering allergic responses.
MA1-5025 detects Fc epsilonR1 alpha (FITC) from human samples. The CRA2 (AER24) monoclonal antibody reacts with the Fc epsilonR1 alpha subunit on a region that overlaps the region of the IgE binding site, thus it competes with IgE for the receptor bindingMA1-5025 has been successfully used in immunocytochemistry, flow cytometry, and immunohistochemistry applications.The MA1-5025 immunogen is region of Fc epsilonR1 alpha which overlaps with the IgE binding site.Specifications
Fc epsilon R1 alpha | |
Monoclonal | |
1mg/mL | |
FCER1A | |
RP11-550P17.3, FCE1A, FcERI | |
Mouse | |
Purified | |
RUO | |
2205 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
CRA2 (AER24) | |
FITC | |
P12319 | |
FCER1A | |
Region of Fc epsilonR1 alpha which overlaps with the IgE binding site. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title