Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcΕRI α subunit Mouse anti-Human, Clone: 3G6, MilliporeSigma™

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma 05491

Catalog No. 05-491-MI


Add to cart

Description

Description

Anti-FcΕRI α subunit Antibody, clone 3G6 detects level of FcΕRI α subunit and has been published and validated for use in immunoprecipitation.
Specifications

Specifications

FcεRI α subunit
3G6
mouse ascites containing 0.05% sodium azide
Ascites
Murine
IgG1
200μL
Primary
Human
Immunoprecipitation
Unconjugated
NM_002001
FCER1A, FcERI, FCE1A
Transfected cells expressing the α-chain of the human high-affinity receptor for immunoglobulin E (FcεRI)
Ascites
−20°C for 24 months.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only