Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/CD32 Mouse anti-Human, Clone: 190723, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen Fc gamma RII/CD32
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB Ala46-Pro217 Accession # P31994
Clone 190723
Applications Western Blot, Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1330SP
View Documents
R&D Systems
MAB1330SP
25 ug
Each for $114.50
Add to cart
 
MAB1330100
View Documents
R&D Systems
MAB1330100
100 ug
Each for $377.00
Add to cart
 
MAB1330500
View Documents
R&D Systems
MAB1330500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Fc gamma RII/CD32 Monoclonal antibody MAB1330 Specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

Fc gamma RII/CD32
190723
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2a
2213
RUO
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB Ala46-Pro217 Accession # P31994
Western Blot, Flow Cytometry, CyTOF
Purified
P31994
Monoclonal
Primary
Detects human Fc gamma RIIA and human Fc gamma RIIB in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Fc gamma RII.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit