Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc fragment of IgA, receptor for, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human FCAR protein.

Manufacturer:  Abnova Corporation H00002204B01

Catalog No. 89-005-353


Add to cart

Description

Description

This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: MDPKQTTLLCLVLCLGQRIQAQEGDFPMPFISAKSSPVIPLDGSVKIQCQAIREAYLTQLMIIKNSTYREIGRRLKFWNETDPEFVIDHMDANKAGRYQCQYRIGHYRFRYSDTLELVVTGLYGKPFLSADRGLVLMPGENISLTCSSAHIPFDRFSLAKEGELSLPQHQSGEHPANFSLGPVDLNVSGIYRCYGWYNRSPYLWSFPSNALELVVTDSIHQDYTTQNLIRMAVAGLVLVALLAILVENWHSHTALNKEASADVAEPSWSQQMCQPGLTFARTPSVCK
Specifications

Specifications

Fc fragment of IgA, receptor for
Unconjugated
No additive
BC027953
CD89
Mouse
50 uL
Membrane Receptors
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human FCAR protein.
FCAR
Serum
FCAR
FCAR (AAH27953, 1 a.a. ~ 287 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2204
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit