Learn More
Fc receptor-like 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FCRL1.
Supplier: Abnova Corporation H00115350A01
Description
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains three extracellular C2-like immunoglobulin domains, a transmembrane domain and a cytoplasmic domain with two immunoreceptor-tyrosine activation motifs. This protein may play a role in the regulation of cancer cell growth. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: ASPSHPTEGSPVTLTCKMPFLQSSDAQFQFCFFRDTRALGPGWSSSPKLQIAAMWKEDTGSYWCEAQTMASKVLRSRRSQINVHRVPVADVSLETSpecifications
Fc receptor-like 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant FCRL1. | |
FCRL1 | |
DKFZp667O1421/FCRH1/IFGP1/IRTA5/RP11-367J7.7 | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_052938 | |
FCRL1 | |
FCRL1 (NP_443170, 23 a.a. ∼ 117 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
115350 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.