Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL4 Mouse anti-Human, Clone: 580810, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524262

Catalog No. PIMA524262


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human FCRL1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein has four extracellular C2-type immunoglobulin domains, a transmembrane domain and a cytoplasmic domain that contains three immune-receptor tyrosine-based inhibitory motifs. This protein may play a role in the function of memory B-cells in the epithelia. Aberrations in the chromosomal region encoding this gene are associated with non-Hodgkin lymphoma and multiple myeloma.
Specifications

Specifications

FCRL4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q96PJ5
CD307d, FCRH4, IGFP2, IRTA1, Fc receptor-like protein 4, IFGP family protein 2, fc receptor homolog 4, fcR-like protein 4, hIFGP2, immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
83417
Flow Cytometry
580810
Unconjugated
FCRL4
Lyophilized
FCRL4
Mouse myeloma cell line NS0-derived recombit human FCRL4/FcRH4 Gln16-Asp385
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit