Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527518

Catalog No. PIPA527518


Add to cart

Description

Description

PA5-27518 targets FEN1 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-27518 immunogen is recombit fragment corresponding to a region within amino acids 169 and 380 of Human FEN1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene removes 5 ft. overhanging flaps in DNA repair and processes the 5 ft. ends of Okazaki fragments in lagging strand DNA synthesis. Direct physical interaction between this protein and AP endonuclease 1 during long-patch base excision repair provides coordinated loading of the proteins onto the substrate, thus passing the substrate from one enzyme to another. The protein is a member of the XPG/RAD2 endonuclease family and is one of ten proteins essential for cell-free DNA replication. DNA secondary structure can inhibit flap processing at certain trinucleotide repeats in a length-dependent manner by concealing the 5 ft. end of the flap that is necessary for both binding and cleavage by the protein encoded by this gene. Therefore, secondary structure can deter the protective function of this protein, leading to site-specific trinucleotide expansions.
Specifications

Specifications

FEN1
Polyclonal
Unconjugated
FEN1
Liquid
FEN1
Recombit fragment corresponding to a region within amino acids 169 and 380 of Human FEN1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.72 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P39748, P39749, Q5XIP6
FEN-1, MF1, RAD2, DNase IV, flap endonuclease 1, hFEN-1, maturation factor 1, maturation factor-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14156, 2237, 84490
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit