Learn More
Fentanyl Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175146
Catalog No. PA175146
Add to cart
Description
Description
PA1-75146 specifically detects Fentanyl. PA1-75146 has been successfully used in ELISA applications. The PA1-75146 immunogen is Fentanyl(p)-BSA.
Fentanyl is a synthetic, lipophilic phenylpiperidine opioid agonist with analgesic and anesthetic properties. Fentanyl selectively binds to the mu-receptor in the central nervous system (CNS) thereby mimicking the effects of endogenous opiates. Stimulation of the mu-subtype opioid receptor stimulates the exchange of GTP for GDP on the G-protein complex and subsequently inhibits adenylate cyclase. This results in a decrease in intracellular cAMP and leads to a reduction in the release of neurotransmitters such as substance P, GABA, dopamine, acetylcholine and noradrenaline. The analgesic effect of fentanyl is likely due to its metabolite morphine, which induces opening of G-protein-coupled inwardly rectifying potassium (GIRK) channels and blocks the opening of N-type voltage-gated calcium channels, thereby resulting in hyperpolarization and reduced neuronal excitability. [National Center for Biotechnology Information. PubChem Compound Database; CID=3345]
Specifications
Specifications
Fentanyl
Polyclonal
Unconjugated
Liquid
Sheep
Ig
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical
ELISA
9.79 mg/mL
PBS with 0.09% sodium azide; pH 7.2
C22H28N2O; Fentanil; Duragesic; Fentanest; Phentanyl
Fentanyl(p)-BSA
Purified
RUO
Antibody
Polyclonal