Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Fer, DyLight 650, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP120089C
Description
Fer Polyclonal specifically detects Fer in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
Fer | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated | |
P16591 | |
FER | |
Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot# P16591] | |
Affinity Purified | |
RUO | |
Primary | |
Human. Predicted to react with mouse and canine based on 100% sequence homology. | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Proto-oncogene c-Fer, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer | |
Rabbit | |
94 kDa | |
0.1 mL | |
Protein Kinase | |
2241 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only