Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FECH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA529733
Description
Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (92%), Rat (93%), Xenopus laevis (87%), Pig (89%), Chicken (85%), Chimpanzee (100%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FECH Polyclonal antibody specifically detects FECH in Human samples. It is validated for Western Blot.Specifications
FECH | |
Polyclonal | |
Unconjugated | |
FECH | |
AI894116; EPP; FCE; fch; Fcl; FECH; ferrochelatase; ferrochelatase, mitochondrial; Heme synthase; heme synthetase; protoheme ferro-lyase; protoporphyria | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.74 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P22830 | |
FECH | |
Recombinant fragment corresponding to a region within amino acids 129 and 370 of Human Ferrochelatase | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction