Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fetuin B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529468

Catalog No. PIPA529468


Add to cart

Description

Description

PA5-29469 targets HERPUD1 in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-29469 immunogen is recombit protein fragment corresponding to a region within amino acids 9 and 251 of Human HERPUD1.

The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.
Specifications

Specifications

Fetuin B
Polyclonal
Unconjugated
FETUB
Liquid
FETUB
Recombit fragment corresponding to a region within amino acids 86 and 361 of Human Fetuin B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Q9UGM5, Q9QXC1
16G2, Gugu, IRL685, fetuin-B, fetuin-like protein IRL685
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26998, 59083
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit