Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fetuin A, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA128320

Catalog No. PIPA128320

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Alpha 2 HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.

PA1-28320 detects Fetuin from various samples.PA1-28320 has been successfully used in Western blot and ELISA applications. The predicted molecular weight is 31 kDa.The PA1-28320 immunogen is: Full length native protein (purified from plasma).
Specifications

Specifications

Fetuin
Polyclonal
Unconjugated
AHSG
Purified
AHSG
Full length native protein (purified from plasma).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
ELISA, Western Blot
1mg/mL
PBS with no preservative
P02765
16G2, Gugu, IRL685
Chicken
IgY
50μg
Store at -20°C
Primary
197
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit