Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEZ1 Mouse, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611710

Catalog No. BDB611710


Add to cart

Description

Description

The development of cancer is a multi-step process involving DNA alterations, oncogene activation, and/or the inactivation/deletion of an anti-onocogene or tumor suppressor. Tumor suppressors are involved in many facets of cell biology,such as cell cycle regulation and development. FEZ1 (F37/Esophageal cancer related gene-coding leucine zipper motif) is a putative DNA-binding protein with homology to cAMP-responsive activating-transcription factor 5 (Atf-5). The structure of FEZ1 includes DNA-binding and leucine zipper domains at amino acids 301-369 , as well as a putative cAMP-dependent phosphorylation site at Ser-29. FEZ1 is ubiquitously expressed in normal tissues with the most abundant expression in testes and brain, but is absent in 31 different cancer cell lines and 16 primary tumors. In addition, the FEZ1 gene has missense mutations in two primary esophageal cancers and a nonsense mutation in a prostate cancer cell line. Several FEZ1-expressing tumors have internally truncated FEZ1 mRNA transcripts. Thus, FEZ1 inactivation in cancers may involve both allelic loss
and point mutations and implicates FEZ1 in cAMP-dependent tumor suppression.

Host Species: Mouse
Clone: 1
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Human FEZ1 aa. 381-592

Immunofluorescence, Western Blotting

Specifications

Specifications

FEZ1
1
Unconjugated
Affinity Purified
Human FEZ1 aa. 381-592
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit